Order Entry
Canada
ContactUsLinkComponent
Cynomolgus Monkey Recombinant ANGPT2 (from HEK293 cells)
Cynomolgus Monkey Recombinant ANGPT2 (from HEK293 cells)
Catalog # 77538-618
Supplier:  Creative Biomart
CAS Number:  
undefined
Cynomolgus Monkey Recombinant ANGPT2 (from HEK293 cells)
Catalog # 77538-618
Supplier:  Creative Biomart
Supplier Number:  ANGPT2-1173C
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Cynomolgus monkey
  • Size:
    50 µg
  • Storage conditions:
    4 °C
  • Endotoxin content:
    <1 EU/μg protein as determined by the LAL method
  • Biological activity:
    Testing in progress
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    angiopoietin 2
  • Protein/peptide name:
    ANGPT2
  • Purity:
    >90% as determined by SDS-PAGE
  • Molecular weight:
    56.2 kDa
  • Endotoxin level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20
  • Cat. no.:
    77538-618

Specifications

About this item