Firefly Recombinant Luciferase (from E. coli)
77540-184
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Firefly
- Size:1 mg
- Storage conditions:4 °C
- Biological activity:2.9×10¹¹ RLU/mg
- Reconstitution instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein/peptide name:Luciferase
- Purity:>95%
- Molecular weight:62 Kda
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77540-184