Human Recombinant LRP6 (from E. coli)
77540-152
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 µg
- Storage conditions:4 °C
- Endotoxin content:<1 EU/μg by LAL
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:low density lipoprotein receptor-related protein 6
- UniProtKB:O75581
- Protein/peptide name:LRP6
- Purity:>90% by SDS-PAGE
- Molecular weight:32.2 kDa
- Sequence:MGSSHHHHHHGLNDIFEAQKIEWHESSGLVPRGSHMASMTGGQQMGRGSEFWIDSRQNMIRKAQEDGSQGFTVVVSSVPSQNLEIQPYDLSIDIYSRYIYWTCEATNVINVTRLDGRSVGVVLKGEQDRPRAVVVNPEKGYMYFTNLQERSPKIERAALDGTEREVLFFSGLSKPIALALDSRLGKLFWADSDLRRIESSDLSGANRIVLEDSNILQPVGLTVFENWLYWIDKQQQMIEKIDMTGREGRTKVQARIAQLSDIHAVKELNLQEYRQHPCAQDNGG
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77540-152