Human Recombinant INS (from E. coli)
Catalog # 77540-026
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:50 µg
- Storage conditions:4 °C
- Reconstitution instructions:Reconstitute to 1 mg/ml in sterile phosphate buffered saline
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:insulin
- UniProtKB:P01308
- Protein/peptide name:INS
- Purity:>99%
- Molecular weight:9394.67 Da
- Sequence:FVNQHLCGSHLVEALYLVCGERGFFYTPKTRREAEDLQVGQVELGGGPGAGSLQPLALEGSLQKRGIVEQCCTS ICSLYQLENYCN
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77540-026