Human Recombinant HLA-G (from HEK293T cells)
Supplier: Creative Biomart
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293T cells
- Species:Human
- Storage conditions:4 °C
- Biological activity:Cell treatment
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:major histocompatibility complex, class I, G
- UniProtKB:P17693
- Protein/peptide name:HLA-G
- Purity:>80% as determined by SDS-PAGE and coomassie blue staining
- Molecular weight:38.3 kDa
- Endotoxin level:Low
- Concentration:>50 μg/ml as determined by microplate BCA method
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Specifications
Frequently Bought Together