Human Recombinant FZD6 (from HEK293 cells)
77539-550
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage conditions:4 °C
- Endotoxin content:<1 EU/μg of the protein as determined by the LAL method
- Reconstitution instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.25 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:frizzled class receptor 6
- UniProtKB:O60353
- Protein/peptide name:FZD6
- Purity:>95% as determined by SDS-PAGE
- Molecular weight:The recombinant human FZD6 consists 146 amino acids and predicts a molecular mass of 17.1 kDa
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-550