Human Recombinant FOXA1 (from E. coli)
Supplier: Creative Biomart
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:4 °C
- Endotoxin content:<1 EU/1 µg (determined by the LAL method)
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:forkhead box A1
- UniProtKB:P55317
- Protein/peptide name:FOXA1
- Purity:≥80% as determined by SDS-PAGE
- Molecular weight:~15 kDa
- Endotoxin level:Low
- Concentration:0.2 mg/ml
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Specifications
Frequently Bought Together