Human Recombinant ERBB3 (from HEK293 cells)
Catalog # 77539-410
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:200 µg
- Storage conditions:4 °C
- Endotoxin content:<1 EU/1 μg (determined by the LAL method)
- Biological activity:Immobilized HER3/ERBB3 protein, human, recombinant (ECD) at 2 μg/ml (100 μl/well) can bind NRG1 Beta 1 Protein, human, recombinant, biotinylated (EGF domain, Fc Tag), the EC50 is 122 ng/ml.
- Reconstitution instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.415 mg/ml. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:ERBB3 v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian)
- UniProtKB:P21860
- Protein/peptide name:ERBB3
- Purity:>85% as determined by SDS-PAGE
- Molecular weight:The recombinant human ERBB3 consists 630 amino acids and predicts a molecular mass of 69.3 kDa
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-410