Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant DHODH (from Mammalian cells)
Mouse Recombinant DHODH (from Mammalian cells)
Supplier:  Creative Biomart
Certificates
undefined
Mouse Recombinant DHODH (from Mammalian cells)
Supplier:  Creative Biomart

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    Mammalian cells
  • Species:
    Mouse
  • Storage conditions:
    4 °C
  • Endotoxin content:
    <1 EU/μg of the protein as determined by the LAL method
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    dihydroorotate dehydrogenase (quinone)
  • UniProtKB:
    O35435
  • Protein/peptide name:
    DHODH
  • Purity:
    >80%
  • Endotoxin level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20

Specifications

Frequently Bought Together