Human Recombinant DDX3Y (from E. Coli or Yeast)
Catalog # 77539-276
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:YeastE. coli
- Species:Human
- Size:50 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:DEAD (Asp-Glu-Ala-Asp) box polypeptide 3, Y-linked
- UniProtKB:O15523
- Protein/peptide name:DDX3Y
- Purity:>90%
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-276