Human Recombinant CXCL12 (from Human cells)
Catalog # 77539-240
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Human cells
- Species:Human
- Size:10 µg
- Storage conditions:4 °C
- Endotoxin content:<1 EU/μg (determined by the LAL method)
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:chemokine (C-X-C motif) ligand 12
- UniProtKB:P61073
- Protein/peptide name:CXCL12
- Purity:>90% by SDS-PAGE
- Endotoxin level:Low
- Concentration:0.35 mg/ml
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-240