Pan troglodytes Recombinant CXCL1 (from E. coli)
Supplier: Creative Biomart
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Pan troglodytes
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:chemokine (C-X-C motif) ligand 1
- UniProtKB:H2QPN5
- Protein/peptide name:CXCL1
- Purity:>85% by SDS-PAGE
- Molecular weight:The protein has a calculated MW of 12.12 kDa
- Sequence:MHHHHHHMAR AALSAAPSNP RLLRVALLLL LLVAAGRRAA GASVATELRC QCLQTLQGIHPKNIQSVNVK SPGPHCAQTE VIATLKNGRK ACLNPASPIV KKIIEMLNSD KSN
- Endotoxin level:Low
- Concentration:0.2 mg/ml
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Specifications
Frequently Bought Together