Human Recombinant CTSE (from HEK293 cells)
77539-210
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:10 µg
- Storage conditions:4 °C
- Endotoxin content:<0.1 ng/μg (1 IEU/μg)
- Reconstitution instructions:Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration <100 µg/ml. Dissolve the lyophilized protein in 1X PBS. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:cathepsin E
- UniProtKB:P14091
- Protein/peptide name:CTSE
- Purity:>95% as determined by SEC-HPLC and reducing SDS-PAGE
- Sequence:SLHRVPLRRHPSLKKKLRARSQLSEFWKSHNLDMIQFTESCSMDQSAKEPLINYLDMEYFGTISIGSPPQNFTVI FDTGSSNLWVPSVYCTSPACKTHSRFQPSQSSTYSQPGQSFSIQYGTGSLSGIIGADQVSVEGLTVVGQQFGESV TEPGQTFVDAEFDGILGLGYPSLAVGGVTPVFDNMMAQNLVDLPMFSVYMSSNPEGGAGSELIFGGYDHSHFSGS LNWVPVTKQAYWQIALDNIQVGGTVMFCSEGCQAIVDTGTSLITGPSDKIKQLQNAIGAAPVDGEYAVECANLNV MPDVTFTINGVPYTLSPTAYTLLDFVDGMQFCSSGFQGLDIHPPAGPLWILGDVFIRQFYSVFDRGNNRVGLAPA VPVDHHHHHH
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-210