Human Recombinant CBR3 (from E. coli)
Supplier: Creative Biomart
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:carbonyl reductase 3
- UniProtKB:O75828
- Protein/peptide name:CBR3
- Purity:>95% by SDS-PAGE
- Endotoxin level:Low
- Concentration:1 mg/ml (Determined by Bradford assay)
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Specifications
Frequently Bought Together