Order Entry
Canada
ContactUsLinkComponent
Human Recombinant BMP2 (from E. coli)
Human Recombinant BMP2 (from E. coli)
Supplier:  Creative Biomart
Certificates
undefined
Human Recombinant BMP2 (from E. coli)
Supplier:  Creative Biomart

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    4 °C
  • Endotoxin content:
    <1 EU/µg of rHuBMP-2 as determined by LAL method
  • Reconstitution instructions:
    We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute in sterile distilled water to a concentration of 0.1 - 1.0 mg/ml. Stock solutions should be apportioned into working aliquots and stored at ≤−20 centigrade. Further dilutions should be made in appropriate buffered solutions. Do not reconstitute in PBS or cell culture media.
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    bone morphogenetic protein 2
  • UniProtKB:
    P12643
  • Protein/peptide name:
    BMP2
  • Purity:
    >95% by SDS-PAGE and HPLC analysis
  • Molecular weight:
    Approximately 26 kDa, a homodimeric protein consisting of two 115 amino acid non-glycosylated polypeptide chains
  • Sequence:
    MQAKHKQRKRLKSSCKRHPLYVDFSDVGWNDWIVAPPGYHAFYCHGECPFPLADHLNSTNHAIVQTLVNSVNSKIPKACCVPTELSAISMLYLDENEKVVLKNYQDMVVEGCGCR
  • Endotoxin level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20

Specifications

Frequently Bought Together