Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant AR (from E. coli)
 
undefined
Human Recombinant AR (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    4 °C
  • Endotoxin content:
    Not determined
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    androgen receptor
  • UniProtKB:
    P10275
  • Protein/peptide name:
    AR
  • Purity:
    >95%
  • Molecular weight:
    62 kDa
  • Endotoxin level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20
  • Tested applications:
    ELISA, western blotting, dot blotting, immunogen, protein array, function assay and more. Before use, heat the WBC control at 95 °C for 5 minutes. Spin down the sample and load 5 µl per lane onto SDS-PAGE gel.

 

Frequently Bought Together