Mouse Recombinant Angpt2 (from CHO cells)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Mouse
- Storage conditions:4 °C
- Endotoxin content:<0.10 EU/μg of the protein by the LAL method
- Biological activity:Measured by its ability to activate Tie-2 in C6 rat glial cells transfected with human Tie-2. 1 μg/ml of recombinant mouse Angiopoietin-2 significantly induces phosphorylation of human Tie-2 in the presence of a cross-linking antibody, mouse anti-polyHistidine monoclonal antibody.
- Reconstitution instructions:Reconstitute at 100 μg/ml in sterile, deionized water
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:angiopoietin 2
- Protein/peptide name:Angpt2
- Purity:>95% by SDS-PAGE under reducing conditions and visualized by silver stain
- Molecular weight:Predicted molecular mass: 49.7 kDa. SDS-PAGE: 60 - 66 kDa, reducing conditions.
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Frequently Bought Together