Order Entry
Canada
ContactUsLinkComponent
Mouse Recombinant Aebp1 (from E. coli)
Mouse Recombinant Aebp1 (from E. coli)
Supplier:  Creative Biomart
Certificates
undefined
Mouse Recombinant Aebp1 (from E. coli)
Supplier:  Creative Biomart

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    4 °C
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    AE binding protein 1
  • UniProtKB:
    Q640N1
  • Protein/peptide name:
    Aebp1
  • Purity:
    >90% (SDS-PAGE)
  • Endotoxin level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20

Specifications

Frequently Bought Together