Human Recombinant ACE2 (from HEK293 cells)
Supplier: Creative Biomart
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:4 °C
- Endotoxin content:<0.1 ng/µg (1 EU/µg) as determined by LAL test
- Biological activity:Loaded 2019-nCoV S protein RBD-mFc on AMC biosensor, can bind human ACE-2-His with an affinity constant of 2.06 nM as determined in BLI assay
- Reconstitution instructions:It is recommended to reconstitute the lyophilized powder in sterile water to 100 - 200 µg/ml, and further dilute with other solvents. The dissolved protein can be stored at 4 °C for 2 - 7 days and should be used up as soon as possible. If not in use for a short time, please store at −20 °C.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:angiotensin I converting enzyme 2
- UniProtKB:Q9BYF1
- Protein/peptide name:ACE2
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:84.63 kDa
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Specifications
Frequently Bought Together