Mouse Native A2m (from Serum)
Catalog # 77538-498
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Native
- Source:Serum
- Species:Mouse
- Size:100 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:Alpha-2-macroglobulin
- Protein/peptide name:A2m
- Purity:>90% by SDS-PAGE
- Endotoxin level:Low
- Concentration:Typically >0.5 mg/ml
- Nuclease-free:No
- Shipping temperature:20
- Cat. no.:77538-498