ContactUsLinkComponent
TF2-HQKLVFFAK-TQ2 [TF2-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Lys-TQ2]
TF2-HQKLVFFAK-TQ2 [TF2-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Lys-TQ2]
Catalog # 77461-434
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
TF2-HQKLVFFAK-TQ2 [TF2-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Lys-TQ2]
Catalog # 77461-434
Supplier:  AAT BIOQUEST INC
Supplier Number:  13206
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    1 mg
  • Protein/peptide name:
    TF2-HQKLVFFAK-TQ2 [TF2-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Lys-TQ2]
  • Cat. no.:
    77461-434
Specifications
About this item
TF2-HQKLVFFAK-TQ2 [TF2-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Lys-TQ2] has much stronger fluoresce upon protease cleavage than the commonly used FRET peptide MCA-HQKLVFFA (K-DNP), a secretase FRET peptide substrate.
  • Stable for 12 months
  • Optimized for FRET applications
Quality tested in-house.
Caution: For research use only.