ContactUsLinkComponent
ReadiView™ Blue Real-Time PCR Visualization Dye *200X*
ReadiView™ Blue Real-Time PCR Visualization Dye *200X*
  77461-618
 :  
undefined
ReadiView™ Blue Real-Time PCR Visualization Dye *200X*
  77461-618
 :  17300
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    ReadiView™ blue real-time PCR visualization dye *200X*
  • Size:
    500 tests
  • Cat. no.:
    77461-618
ReadiView™ Blue Real-Time PCR Visualization Dye is a 200X concentrated, ready-to-use, inert dye that enhances the visibility of real-time PCR reactions for more accurate pipetting and plate loading.
  • Stable for 12 months
Quality tested in-house.
 : For research use only.