ContactUsLinkComponent
iFluor® 790-Wheat Germ Agglutinin (WGA) Conjugate
iFluor® 790-Wheat Germ Agglutinin (WGA) Conjugate
Catalog # 77462-036
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
iFluor® 790-Wheat Germ Agglutinin (WGA) Conjugate
Catalog # 77462-036
Supplier:  AAT BIOQUEST INC
Supplier Number:  25563
CAS Number:  
Specifications
  • Description:
    iFluor® 790-Wheat germ agglutinin (WGA) conjugate
  • Size:
    1 mg
  • Cat. no.:
    77462-036
Specifications
About this item
iFluor® 790 labeled WGA conjugate exhibits a distinct near-infrared fluorescence characteristic, emitting maximally at 812 nm.
  • Stable for 12 months
Wheat germ agglutinin (WGA) is a well-studied lectin known for its binding affinity to N-acetyl-D-glucosamine and sialic acid, making it a valuable tool in various biological applications.
Caution: For research use only.