ContactUsLinkComponent
Gelite™ Safe DNA Gel Stain *GelRed Replacement, 10000X in Water*
Gelite™ Safe DNA Gel Stain *GelRed Replacement, 10000X in Water*
Catalog # 77461-662
Supplier:  AAT BIOQUEST INC
CAS Number:  
undefined
Gelite™ Safe DNA Gel Stain *GelRed Replacement, 10000X in Water*
Catalog # 77461-662
Supplier:  AAT BIOQUEST INC
Supplier Number:  17708
CAS Number:  
Specifications
  • Description:
    Gelite™ safe DNA gel stain *GelRed replacement, 10000X in water*
  • Size:
    0.5 ml
  • Cat. no.:
    77461-662
Specifications
About this item
Gelite™ Safe has been developed specifically to be less hazardous than EtBr for staining DNA in agarose and acrylamide gels with much higher sensitivity.
  • Stable for 12 months
AAT Bioquest is committed to designing our products to be environment-friendly.
Caution: For research use only.