Human Recombinant MTOR (from E. coli)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:mechanistic target of rapamycin (serine/threonine kinase)
- UniProtKB:P42345
- Protein/peptide name:MTOR
- Purity:>80% by SDS-PAGE
- Molecular weight:38.4 kDa
- Sequence:MSPILGYWKIKGLVQPTRLLLEYLEEKYEEHLYERDEGDKWRNKKFELGLEFPNLPYYIDGDVKLTQSMAIIRYIADKHNMLGGCPKERAEISMLEGAVLDIRYGVSRIAYSKDFETLKVDFLSKLPEMLKMFEDRLCHKTYLNGDHVTHPDFMLYDALDVVLYMDPMCLDAFPKLVCFKKRIEAIPQIDKYLKSSKYIAWPLQGWQATFGGGDHPPKSDLVPRGSELIRVAILWHEMWHEGLEEASRLYFGERNVKGMFEVLEPLHAMMERGPQTLKETSFNQAYGRDLMEAQEWCRKYMKSGNVKDLTQAWDLYYHVFRRISKQ
- Endotoxin level:Low
- Concentration:2.5 mg/ml
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Frequently Bought Together