Mouse Recombinant IMPDH2 (from Mammalian cells)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Mammalian cells
- Species:Mouse
- Storage conditions:4 °C
- Endotoxin content:<1.0 EU/µg of the protein as determined by the LAL method
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:IMP (inosine 5-monophosphate) dehydrogenase 2
- UniProtKB:P24547
- Protein/peptide name:IMPDH2
- Purity:>80%
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Frequently Bought Together