Human Recombinant IL8 (from CHO cells)
Catalog # 77539-996
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Size:10 µg
- Storage conditions:4 °C
- Biological activity:Shows the biological function of the IL-8 moiety and exerts a prolonged circulating half-life caused by the modified Fc domain
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:Barley yellow dwarf virus - PAV
- UniProtKB:P10145
- Protein/peptide name:IL8
- Purity:≥98% (SDS-PAGE)
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-996