Order Entry
Canada
ContactUsLinkComponent
Rat Recombinant Il18 (from E. coli)
Rat Recombinant Il18 (from E. coli)
Catalog # 77539-916
Supplier:  Creative Biomart
CAS Number:  
undefined
Rat Recombinant Il18 (from E. coli)
Catalog # 77539-916
Supplier:  Creative Biomart
Supplier Number:  IL18-522R
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Size:
    50 µg
  • Storage conditions:
    4 °C
  • Reconstitution instructions:
    Reconstitute in water to a concentration of 0.1 - 1.0 µg/µl. Gentle pipetting to mix it well. The solution can then be diluted into other aqueous buffers and stored at 4 °C for 1 week or −20 °C for future use.
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    interleukin 18
  • UniProtKB:
    P97636
  • Protein/peptide name:
    Il18
  • Purity:
    >95% by SDS-PAGEand HPLC analysis
  • Endotoxin level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20
  • Cat. no.:
    77539-916

Specifications

About this item