Human Recombinant HSP90AA1 (from E. coli)
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:heat shock protein 90 kDa alpha (cytosolic), class A member 1
- UniProtKB:Q86SX1
- Protein/peptide name:HSP90AA1
- Purity:>90% by SDS-PAGE
- Endotoxin level:Low
- Concentration:1 mg/ml (determined by Bradford assay)
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Frequently Bought Together