Order Entry
Canada
ContactUsLinkComponent
Human Papilloma Virus 52 (HPV-52) Recombinant HPV52-L1 (from E. coli)
Human Papilloma Virus 52 (HPV-52) Recombinant HPV52-L1 (from E. coli)
Catalog # 77539-788
Supplier:  Creative Biomart
CAS Number:  
undefined
Human Papilloma Virus 52 (HPV-52) Recombinant HPV52-L1 (from E. coli)
Catalog # 77539-788
Supplier:  Creative Biomart
Supplier Number:  HPV52-L1-06H
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human papilloma virus 52 (HPV-52)
  • Size:
    25 µg
  • Storage conditions:
    4 °C
  • Endotoxin content:
    <1.0 EU/µg by LAL method
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    L1
  • UniProtKB:
    Q05138
  • Protein/peptide name:
    HPV52-L1
  • Purity:
    >85% by SDS-PAGE
  • Molecular weight:
    59.12 kDa
  • Sequence:
    MNHKVHHHHH HMQMSVWRPS EATVYLPPVP VSKVVSTDEY VSRTSIYYYA GSSRLLTVGHPYFSIKNTSS GNGKKVLVPK VSGLQYRVFR IKLPDPNKFG FPDTSFYNPE TQRLVWACTGLEIGRGQPLG VGISGHPLLN KFDDTETSNK YAGKPGIDNR ECLSMDYKQT QLCILGCKPPIGEHWGKGTP CNNNSGNPGD CPPLQLINSV IQDGDMVDTG FGCMDFNTLQ ASKSDVPIDICSSVCKYPDY LQMASEPYGDSLFFFLRREQMFVRHFFNRAGTLGDPVPGDLYIQGSNSGNTATVQSSAFF PTPSGSMVTS ESQLFNKPYW LQRAQGHNNG ICWGNQLFVT VVDTTRSTNMTLCAEVKKES TYKNENFKEY LRHGEEFDLQ FIFQLCKITL TADVMTYIHK MDATILEDWQFGLTPPPSAS LEDTYRFVTS TAITCQKNTP PKGKEDPLKD YMFWEVDLKE KFSADLDQFPLGRKFLLQAG LQARPKLKRP ASSAPRTSTK KKKVKRHHHH HHHH
  • Endotoxin level:
    Low
  • Concentration:
    0.4 mg/ml
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20
  • Cat. no.:
    77539-788

Specifications

About this item