Human Recombinant HNRNPK (from E. coli)
Catalog # 77539-780
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:heterogeneous nuclear ribonucleoprotein K
- UniProtKB:P61978
- Protein/peptide name:HNRNPK
- Purity:>95% by SDS-PAGE
- Molecular weight:33 kDa (299aa), confirmed by MALDI-TOF
- Sequence:MGSSHHHHHH SSGLVPRGSH MGSMETEQPE ETFPNTETNG EFGKRPAEDM EEEQAFKRSR NTDEMVELRI LLQSKNAGAV IGKGGKNIKA LRTDYNASVS VPDSSGPERI LSISADIETI GEILKKIIPT LEEGLQLPSP TATSQLPLES DAVECLNYQH YKGSDFDCEL RLLIHQSLAG GIIGVKGAKI KELRENTQTT IKLFQECCPH STDRVVLIGG KPDRVVECIK IILDLISESP IKGRAQPYDP NFYDETYDYG GFTMMFDDRR GRPVGFPMRG RGGFDRMPPG RGGRPMPPS
- Endotoxin level:Low
- Concentration:1 mg/ml (determined by Bradford assay)
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-780