Whale Recombinant HB (from E. coli)
Supplier: Creative Biomart
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Whale
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:Asparagus virus 2
- Protein/peptide name:HB
- Purity:>90% by SDS-PAGE
- Molecular weight:HBA: 16.2 kDa, HBB: 18.2 kDa
- Sequence:HBA: MVLSPADKTNVKAAWAKVGNHAADFGAEALERMFMSFPSTKTYFSHFDLGHNSTQVKGHGKKVADALTKAVGHLDTLPDALSDLSDLHAHKLRVDPVNFKLLSHCLLVTLAAHLPGDFTPSVHASLDKFLASVSTVLTSKYRHHHHHH HBB: MVHLTGEEKSGLTALWAKVNVEEIGGEALGRLLVVYPWTQRFFEHFGDLSTADAVMKNPKVKKHGQKVLASFGEGLKHLDNLKGTFATLSELHCDKLHVDPENFRLLGNVLVVVLARHFGKEFTPELQTAYQKVVAGVANALAHKYHWSHPQFEKHHHHHH
- Endotoxin level:Low
- Concentration:2.5 mg/ml (A280; Abs 0.1% (=1 g/L): 0.86)
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Specifications
Frequently Bought Together