Human Recombinant GUCY2C (from Human cells)
Catalog # 77539-652
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Human cells
- Species:Human
- Size:25 µg
- Storage conditions:4 °C
- Endotoxin content:<1.0 EU/ml (determined by the LAL method)
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:guanylate cyclase 2C (heat stable enterotoxin receptor)
- UniProtKB:P25092
- Protein/peptide name:GUCY2C
- Purity:>90%, by SDS-PAGE
- Sequence:SQVSQNCHNGSYEISVLMMGNSAFAEPLKNLEDAVNEGLEIVRGRLQNAGLNVTVNATFMYSDGLIHNSGDCRSSTCEGLDLLRKISNAQRMGCVLIGPSCTYSTFQMYLDTELSYPMISAGSFGLSCDYKETLTRLMSPARKLMYFLVNFWKTNDLPFKTYSWSTSYVYKNGTETEDCFWYLNALEASVSYFSHELGFKVVLRQDKEFQDILMDHNRKSNVIIMCGGPEFLYKLKGDRAVAEDIVIILVDLFNDQYFEDNVTAPDYMKNVLVLTLSPGNSLLNSSFSRNLSPTKRDFALAYLNGILLFGHMLKIFLENGENITTPKFAHAFRNLTFEGYDGPVTLDDWGDVDSTMVLLYTSVDTKKYKVLLTYDTHVNKTYPVDMSPTFTWKNSKLPNDITGRGPQHHHHHHHHHH
- Endotoxin level:Low
- Concentration:1.0 mg/ml
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-652