Human Recombinant GDF15 (from CHO cells)
77539-590
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:CHO cells
- Size:100 µg
- Storage conditions:4 °C
- Endotoxin content:<1.0 EU/µg as determined by the LAL method
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:growth differentiation factor 15
- UniProtKB:Q99988
- Protein/peptide name:GDF15
- Purity:>97% by SDS-PAGE and analyzed by silver stain
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-590