Human Recombinant GAD67 (from Insect cells)
Catalog # 77539-562
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Insect cells
- Species:Human
- Size:50 µg
- Storage conditions:4 °C
- Endotoxin content:<1.0 EU/µg of the protein as determined by the LAL method
- Reconstitution instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein/peptide name:GAD67
- Purity:>80% as determined by SDS-PAGE
- Molecular weight:The recombinant human GAD1 consists of 605 amino acids and predicts a molecular mass of 68.3 kDa. It migrates as an approximately 64 kDa band in SDS-PAGE in SDS-PAGE under reducing conditions.
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-562