Human Papilloma Virus 6 (HPV-6) Recombinant E6 (from E. coli)
77539-334
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human papilloma virus 6 (HPV-6)
- Size:100 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:E6 protein
- Protein/peptide name:E6
- Purity:>95% by SDS-PAGE
- Molecular weight:The protein has a calculated MW of 18 kDa
- Sequence:MESANASTSATTIDQLCKTFNLSMHTLQINCVFCKNALTTAEIYSYAYKHLKVLFRGGYPYAACACCLEFHGKINQYRHFDYAGYATTVEEETKQDILDVLIRCYLCHKPLCEVEKVKHILTKARFIKLNCTWKGRCLHCWTTCMEDMLPLEHHHHHH
- Endotoxin level:Low
- Concentration:1 mg/ml by BCA
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-334