Human Recombinant DUSP1 (from Wheat Germ)
77539-322
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Wheat germ
- Size:2 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:dual specificity phosphatase 1
- UniProtKB:P28562
- Protein/peptide name:DUSP1
- Purity:Proprietary purification
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-322