Human Recombinant DEFB4 (from E. coli)
77539-282
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Size:10 µg
- Storage conditions:4 °C
- Endotoxin content:<1 EU/µg of rHuBD-4 as determined by LAL method
- Biological activity:Fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration range of 0.1 - 100.0 ng/ml.
- Reconstitution instructions:We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1 - 1.0 mg/ml. Stock solutions should be apportioned into working aliquots and stored at ≤−20 °C. Further dilutions should be made in appropriate buffered solutions.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:defensin beta 4
- UniProtKB:Q496I2
- Protein/peptide name:DEFB4
- Purity:>98% by SDS-PAGE and HPLC analyses
- Molecular weight:Approximately 6.0 kDa, a single non-glycosylated polypeptide chain containing 50 amino acids
- Sequence:EFELDRICGY GTARCRKKCR SQEYRIGRCP NTYACCLRKW DESLLNRTKP
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-282