Order Entry
Canada
ContactUsLinkComponent
Human Recombinant CXCL9
Human Recombinant CXCL9
  77539-246
 :  
undefined
Human Recombinant CXCL9
  77539-246
 :  CXCL9-01H
 :  

 

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Size:
    25 µg
  • Storage conditions:
    4 °C
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    chemokine (C-X-C motif) ligand 9
  • Protein/peptide name:
    CXCL9
  • Endotoxin level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20
  • Cat. no.:
    77539-246