Recombinant Protein A (from E. coli)
Catalog # 77539-130
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Size:1 mg
- Storage conditions:4 °C
- Endotoxin content:<0.01 ng/µg Recombinant Protein A
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:Protein A
- Protein/peptide name:Protein A
- Purity:>95% as determined by SDS-PAGE of 6 μg reduced recombinant proteins. All lots are >95% pure.
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-130