Order Entry
Canada
ContactUsLinkComponent
Recombinant COL10A1
Recombinant COL10A1
Catalog # 77539-110
Supplier:  Creative Biomart
CAS Number:  
undefined
Recombinant COL10A1
Catalog # 77539-110
Supplier:  Creative Biomart
Supplier Number:  COLLAGENX-01H
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Size:
    100 µg
  • Storage conditions:
    4 °C
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    collagen, type X, alpha 1
  • Protein/peptide name:
    COL10A1
  • Endotoxin level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20
  • Cat. no.:
    77539-110

Specifications

About this item