Human Recombinant COL15A1 (from HEK293 cells)
Supplier: Creative Biomart
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:4 °C
- Endotoxin content:<0.10 EU/µg of the protein by the LAL method
- Biological activity:Measured by its ability to enhance neurite outgrowth of E16-E18 rat embryonic cortical neurons. Recombinant Human Collagen XV alpha 1/COL15A1, immobilized at 0.5 μg/ml, is able to significantly enhance neurite outgrowth.
- Reconstitution instructions:Reconstitute at 500 μg/ml in PBS
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:collagen, type X, alpha 1
- UniProtKB:P39059
- Protein/peptide name:COL15A1
- Purity:>95% by SDS-PAGE visualized with silver staining and quantitative densitometry by Coomassie® blue staining
- Molecular weight:29 kDa
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Specifications
Frequently Bought Together