Salmonella enterica Recombinant CdtB (from E. coli)
Catalog # 77539-034
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Salmonella enterica
- Size:25 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:Cytolethal distending toxin B
- UniProtKB:A0A3S4EX57
- Protein/peptide name:CdtB
- Purity:>90% by SDS-PAGE
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77539-034