Order Entry
Canada
ContactUsLinkComponent
Human Recombinant CD3E and CD3D
Human Recombinant CD3E and CD3D
Supplier:  Creative Biomart
Certificates
undefined
Human Recombinant CD3E and CD3D
Supplier:  Creative Biomart

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    4 °C
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    CD3 delta and CD3 epsilon
  • Protein/peptide name:
    CD3E and CD3D
  • Endotoxin level:
    Low
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20

Specifications

Frequently Bought Together