Human Recombinant C9 (from Wheat Germ)
Catalog # 77538-804
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Wheat germ
- Species:Human
- Size:2 µg
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:complement component 9
- UniProtKB:P02748
- Protein/peptide name:C9
- Molecular weight:84.92 kDa
- Sequence:QYTTSYDPELTESSGSASHIDCRMSPWSEWSQCDPCLRQMFRSRSIEVFGQFNGKRCTDAVGDRRQCVPTEPCED AEDDCGNDFQCSTGRCIKMRLRCNGDNDCGDFSDEDDCESEPRPPCRDRVVEESELARTAGYGINILGMDPLSTP FDNEFYNGLCNRDRDGNTLTYYRRPWNVASLIYETKGEKNFRTEHYEEQIEAFKSIIQEKTSNFNAAISLKFTPT ETNKAEQCCEETASSISLHGKGSFRFSYSKNETYQLFLSYSSKKEKMFLHVKGEIHLGRFVMRNRDVVLTTTFVD DIKALPTTYEKGEYFAFLETYGTHYSSSGSLGGLYELIYVLDKASMKRKGVELKDIKRCLGYHLDVSLAFSEISV GAEFNKDDCVKRGEGRAVNITSENLIDDVVSLIRGGTRKYAFELKEKLLRGTVIDVTDFVNWASSINDAPVLISQ KLSPIYNLVPVKMKNAHLKKQNLERAIEDYINEFSVRKCHTCQNGGTVILMDGKCLCACPFKFEGIACEISKQKI SEGLPALEFPNEK
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Tested applications:Enzyme-Linked Immunoabsorbent Assay, western blot (recombinant protein), antibody production, protein array
- Cat. no.:77538-804