Order Entry
Canada
ContactUsLinkComponent
 
Rat Recombinant AR (from E. coli)
 
undefined
Rat Recombinant AR (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Rat
  • Storage conditions:
    4 °C
  • Endotoxin-free:
    No
  • Carrier-free:
    No
  • Protease-free:
    No
  • Animal-free:
    No
  • Protein synonyms:
    androgen receptor
  • UniProtKB:
    P15207
  • Protein/peptide name:
    AR
  • Purity:
    >90% by SDS-PAGE
  • Molecular weight:
    The protein has a calculated MW of 61 kDa
  • Sequence:
    MHHHHHHLRMEVQLGLGRVYPRPPSKTYRGAFQNLFQSVREAIQNPGPRHPEAASIAPPGACLQQRQETSPRRRRRQQHPEDGSPQAHIRGTTGYLALEEEQQPSQQQSASEGHPESGCLPEPGAATAPGKGLPQQPPAPPDQDDSAAPSTLSLLGPTFPGLSSCSADIKDILSEAGTMQLLQQQQQQQQQQQQQQQQQQQQQQEVISEGSSSVRAREATGAPSSSKDSYLGGNSTISDSAKELCKAVSVSMGLGVEALEHLSPGEQLRGDCMYASLLGGPPAVRPTPCAPLAECKGLSLDEGPGKGTEETAEYSSFKGGYAKGLEGESLGCSGSSEAGSSGTLEIPSSLSLYKSGAVDEAAAYQNRDYYNFPLALSGPPHPPPPTHPHARIKLENPSDYGSAWAAAAAQCRYGDLASLHGGSVAGPSTGSPPATASSSWHTLFTAEEGQLYGPGGGGGSSSPSDAGPVAPYGYTRPPQGLASQEGDFSASEVWYPGGVVNRVPYPSPSCVKSEMGPWMENYSGPYGDMRLDSTRDHVLPIDYYFPPQKTCLICGDEASGCHYGALTCGSCKVFFKRA
  • Endotoxin level:
    Low
  • Concentration:
    0.5 mg/ml by BCA
  • Nuclease-free:
    No
  • Grade:
    Ultrapure grade
  • Shipping temperature:
    20

 

Frequently Bought Together