Human Recombinant ALB (from Rice grain)
Supplier: Creative Biomart
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:Rice grain
- Species:Human
- Storage conditions:4 °C
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:Yes
- Protein synonyms:albumin
- UniProtKB:P02768
- Protein/peptide name:ALB
- Purity:>97% as determined by: (a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Tested applications:Formulation of protein therapeutics, Cell Storage: Cryopreservation; Vaccine formulation and manufacturing, development of mammalian cell cultures, infertility treatments, coating for medical devices, drug delivery, in vivo diagnostics
Specifications
Frequently Bought Together