Recombinant Human LRRC15 (from HEK293 cells)
77540-154
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:100 µg
- Storage conditions:4 °C
- Endotoxin content:<1.0 EU/μg of the protein as determined by the LAL method
- Reconstitution instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.25 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:leucine rich repeat containing 15
- UniProtKB:Q8TF66
- Protein/peptide name:LRRC15
- Purity:>85% as determined by SDS-PAGE
- Molecular weight:The recombinant human LRRC15 consists 755 amino acids and predicts a molecular mass of 84.5 kDa.
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77540-154