Recombinant Mouse Kl (from CHO cells)
Catalog # 77540-060
Supplier: Creative Biomart
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Mouse
- Size:50 µg
- Storage conditions:4 °C
- Endotoxin content:<0.01 EU/μg of the protein by the LAL method
- Biological activity:Measured in a cell proliferation assay using BaF3 mouse pro‑B cells transfected with human FGF RIIIc. The ED50 for this effect is typically 0.04 - 0.4 μg/ml in the presence of 1 µg/ml of Recombinant human FGF‑23 and 10 μg/ml of heparin.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:klotho
- UniProtKB:O35082
- Protein/peptide name:Kl
- Purity:>90%, by SDS-PAGE under reducing conditions and visualized by silver stain
- Molecular weight:Predicted molecular mass: 109.5 kDa, SDS-PAGE: 125 - 130 kDa, reducing conditions.
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
- Cat. no.:77540-060