Recombinant Human JAG1 (from HEK293 cells)
Supplier: Creative Biomart
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:4 °C
- Endotoxin content:<1.0 EU/μg protein as determined by the LAL method
- Biological activity:Measured by the ability of the immobilized protein to enhance BMP2-induced alkaline phosphatase activity in C3H10T1/2 mouse embryonic fibroblast cells. The ED50 for this effect is typically 5 - 30 μg/ml.
- Reconstitution instructions:It is recommended that sterile water be added to the vial to prepare a stock solution of 0.2 µg/µl. Centrifuge the vial at 4 °C before opening to recover the entire contents.
- Endotoxin-free:No
- Carrier-free:No
- Protease-free:No
- Animal-free:No
- Protein synonyms:jagged 1
- UniProtKB:P78504
- Protein/peptide name:JAG1
- Purity:>85% as determined by SDS-PAGE
- Molecular weight:The recombinant human JAG1/Fc is a disulfide-linked homodimer. The reduced monomer consists of 1254 amino acids and has a predicted molecular mass of 137 kDa. In SDS-PAGE under reducing conditions, the apparent molecular mass of human JAG1/Fc monomer is approximately 185.5 kDa due to glycosylation.
- Endotoxin level:Low
- Nuclease-free:No
- Grade:Ultrapure grade
- Shipping temperature:20
Specifications
Frequently Bought Together